missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylaniline 98.0+%, TCI America™

Catalog No. B19905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19905G 5 g
B199025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19905G Supplier TCI America Supplier No. B19905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6933-10-4
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00007828
InChI Key MMEGELSFOYDPQW-UHFFFAOYSA-N
Synonym 4-bromo-m-toluidine, 5-amino-2-bromotoluene, 3-methyl-4-bromoaniline, benzenamine, 4-bromo-3-methyl, 4-bromo-3-methyl-phenylamine, 4-bromo-3-methyl-aniline, 4-bromo-3-methylbenzenamine, 4-bromo-3-methylphenylamine, pubchem4339, 4-bromo3-methylaniline
PubChem CID 23359
IUPAC Name 4-bromo-3-methylaniline
SMILES CC1=C(C=CC(=C1)N)Br

Specifications

Melting Point 81°C
Boiling Point 240°C
Quantity 5 g
Formula Weight 186.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-methylaniline