missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methoxyaniline, 97+%

Catalog No. p-7028303
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2554106 5 g
AAB2554114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2554106 Supplier Thermo Scientific Chemicals Supplier No. B2554106
Only null left

CAS: 19056-40-7 | C7H8BrNO | 202.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19056-40-7
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.05
MDL Number MFCD05664063
InChI Key RUTNWXBHRAIQSP-UHFFFAOYSA-N
Synonym 5-amino-2-bromoanisole, 4-bromo-m-anisidine, 4-bromo-3-methoxy-phenylamine, 4-bromo-3-methoxy-aniline, 3-methoxy-4-bromoaniline, 4-bromo-3-methoxy aniline, 4-bromo-3-methoxyphenylamine, benzenamine, 4-bromo-3-methoxy, 4-bromo-3-methoxybenzenamine, pubchem3794
PubChem CID 7018254
IUPAC Name 4-bromo-3-methoxyaniline
SMILES COC1=C(Br)C=CC(N)=C1

Specifications

Melting Point 96°C to 100°C
Quantity 5 g
UN Number UN2811
Beilstein 2690835
Formula Weight 202.05
Percent Purity ≥97%
Chemical Name or Material 4-Bromo-3-methoxyaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only