Learn More
4-Bromo-3-methoxyaniline, 97%
CAS: 19056-40-7 | C7H8BrNO | 202.05 g/mol
$73.61 - $73.61
Chemical Identifiers
| CAS | 19056-40-7 |
|---|---|
| Molecular Formula | C7H8BrNO |
| Molecular Weight (g/mol) | 202.05 |
| MDL Number | MFCD05664063 |
| InChI Key | RUTNWXBHRAIQSP-UHFFFAOYSA-N |
| Synonym | 5-amino-2-bromoanisole, 4-bromo-m-anisidine, 4-bromo-3-methoxy-phenylamine, 4-bromo-3-methoxy-aniline, 3-methoxy-4-bromoaniline, 4-bromo-3-methoxy aniline, 4-bromo-3-methoxyphenylamine, benzenamine, 4-bromo-3-methoxy, 4-bromo-3-methoxybenzenamine, pubchem3794 |
| PubChem CID | 7018254 |
| IUPAC Name | 4-bromo-3-methoxyaniline |
| SMILES | COC1=C(Br)C=CC(N)=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC399810010
|
Thermo Scientific Chemicals
399810010 |
1 g | Glass bottle |
Each for $73.61
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 19056-40-7 | |
| 202.05 | |
| RUTNWXBHRAIQSP-UHFFFAOYSA-N | |
| 7018254 | |
| COC1=C(Br)C=CC(N)=C1 |
| C7H8BrNO | |
| MFCD05664063 | |
| 5-amino-2-bromoanisole, 4-bromo-m-anisidine, 4-bromo-3-methoxy-phenylamine, 4-bromo-3-methoxy-aniline, 3-methoxy-4-bromoaniline, 4-bromo-3-methoxy aniline, 4-bromo-3-methoxyphenylamine, benzenamine, 4-bromo-3-methoxy, 4-bromo-3-methoxybenzenamine, pubchem3794 | |
| 4-bromo-3-methoxyaniline |
Specifications
| 19056-40-7 | |
| 96% min. (GC) | |
| C7H8BrNO | |
| MFCD05664063 | |
| 5-amino-2-bromoanisole, 4-bromo-m-anisidine, 4-bromo-3-methoxy-phenylamine, 4-bromo-3-methoxy-aniline, 3-methoxy-4-bromoaniline, 4-bromo-3-methoxy aniline, 4-bromo-3-methoxyphenylamine, benzenamine, 4-bromo-3-methoxy, 4-bromo-3-methoxybenzenamine, pubchem3794 | |
| COC1=C(Br)C=CC(N)=C1 | |
| 202.05 | |
| 202.05 | |
| 4-Bromo-3-methoxyaniline |
| 96.0°C to 100.0°C | |
| Glass bottle | |
| NH2C6H3(Br)OCH3 | |
| 1 g | |
| RUTNWXBHRAIQSP-UHFFFAOYSA-N | |
| 4-bromo-3-methoxyaniline | |
| 7018254 | |
| 97% |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Warning
RUO – Research Use Only