missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorotoluene 94.0+%, TCI America™

Catalog No. B19355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19355G 5 g
B193525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19355G Supplier TCI America Supplier No. B19355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 452-74-4
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00040940
InChI Key SLFNGVGRINFJLK-UHFFFAOYSA-N
Synonym 4-bromo-3-fluorotoluene, 3-fluoro-4-bromotoluene, benzene, 1-bromo-2-fluoro-4-methyl, 3-fluoro-4-bromo-toluene, 4-bromo-3-fluoro-1-methylbenzene, 1-bromo-2-fluoro-4-methyl-benzene, 3-fluoro-4-bromo-toluone, 1-brom-2-fluor-4-methylbenzol, zlchem 562
PubChem CID 573280
IUPAC Name 1-bromo-2-fluoro-4-methylbenzene
SMILES CC1=CC(=C(C=C1)Br)F

Specifications

Color Yellow
Boiling Point 191°C
Quantity 5 g
UN Number 1993
Formula Weight 189.03
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-3-fluorotoluene