Learn More
4-Bromo-3-fluorotoluene, 98%
CAS: 452-74-4 | C7H6BrF | 189.027 g/mol
$181.82 - $370.71
Chemical Identifiers
| CAS | 452-74-4 |
|---|---|
| Molecular Formula | C7H6BrF |
| Molecular Weight (g/mol) | 189.027 |
| MDL Number | MFCD00040940 |
| InChI Key | SLFNGVGRINFJLK-UHFFFAOYSA-N |
| Synonym | 4-bromo-3-fluorotoluene, 3-fluoro-4-bromotoluene, benzene, 1-bromo-2-fluoro-4-methyl, 3-fluoro-4-bromo-toluene, 4-bromo-3-fluoro-1-methylbenzene, 1-bromo-2-fluoro-4-methyl-benzene, 3-fluoro-4-bromo-toluone, 1-brom-2-fluor-4-methylbenzol, zlchem 562 |
| PubChem CID | 573280 |
| IUPAC Name | 1-bromo-2-fluoro-4-methylbenzene |
| SMILES | CC1=CC(=C(C=C1)Br)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1914209
|
Thermo Scientific Chemicals
A1914209 |
10 g |
Each for $181.82
|
|
|||||
|
AAA1914214
|
Thermo Scientific Chemicals
A1914214 |
25 g |
Each for $370.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 452-74-4 | |
| 189.027 | |
| SLFNGVGRINFJLK-UHFFFAOYSA-N | |
| 573280 | |
| CC1=CC(=C(C=C1)Br)F |
| C7H6BrF | |
| MFCD00040940 | |
| 4-bromo-3-fluorotoluene, 3-fluoro-4-bromotoluene, benzene, 1-bromo-2-fluoro-4-methyl, 3-fluoro-4-bromo-toluene, 4-bromo-3-fluoro-1-methylbenzene, 1-bromo-2-fluoro-4-methyl-benzene, 3-fluoro-4-bromo-toluone, 1-brom-2-fluor-4-methylbenzol, zlchem 562 | |
| 1-bromo-2-fluoro-4-methylbenzene |
Specifications
| 452-74-4 | |
| 187°C to 188°C | |
| C7H6BrF | |
| MFCD00040940 | |
| 2500154 | |
| SLFNGVGRINFJLK-UHFFFAOYSA-N | |
| 1-bromo-2-fluoro-4-methylbenzene | |
| 573280 | |
| 98% |
| 1.494 | |
| 74°C (165°F) | |
| 1.531 | |
| 10 g | |
| 4-bromo-3-fluorotoluene, 3-fluoro-4-bromotoluene, benzene, 1-bromo-2-fluoro-4-methyl, 3-fluoro-4-bromo-toluene, 4-bromo-3-fluoro-1-methylbenzene, 1-bromo-2-fluoro-4-methyl-benzene, 3-fluoro-4-bromo-toluone, 1-brom-2-fluor-4-methylbenzol, zlchem 562 | |
| CC1=CC(=C(C=C1)Br)F | |
| 189.027 | |
| 189.03 | |
| 4-Bromo-3-fluorotoluene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only