missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorotoluene, 98%

Catalog No. AAA1914209
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1914209 10 g
AAA1914214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1914209 Supplier Thermo Scientific Chemicals Supplier No. A1914209
Only null left

CAS: 452-74-4 | C7H6BrF | 189.027 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 452-74-4
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00040940
InChI Key SLFNGVGRINFJLK-UHFFFAOYSA-N
Synonym 4-bromo-3-fluorotoluene, 3-fluoro-4-bromotoluene, benzene, 1-bromo-2-fluoro-4-methyl, 3-fluoro-4-bromo-toluene, 4-bromo-3-fluoro-1-methylbenzene, 1-bromo-2-fluoro-4-methyl-benzene, 3-fluoro-4-bromo-toluone, 1-brom-2-fluor-4-methylbenzol, zlchem 562
PubChem CID 573280
IUPAC Name 1-bromo-2-fluoro-4-methylbenzene
SMILES CC1=CC(=C(C=C1)Br)F

Specifications

Density 1.494
Boiling Point 187°C to 188°C
Flash Point 74°C (165°F)
Refractive Index 1.531
Quantity 10 g
Beilstein 2500154
Formula Weight 189.03
Percent Purity 98%
Chemical Name or Material 4-Bromo-3-fluorotoluene
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only