missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3-fluorophenol 98.0+%, TCI America™
$242.33 - $728.84
Chemical Identifiers
| CAS | 121219-03-2 |
|---|---|
| Molecular Formula | C6H4BrFO |
| Molecular Weight (g/mol) | 190.999 |
| MDL Number | MFCD00051907 |
| InChI Key | MRQYTJXVULSNIS-UHFFFAOYSA-N |
| Synonym | 3-fluoro-4-bromophenol, 4-bromo-3-fluoro-phenol, phenol, 4-bromo-3-fluoro, pubchem1489, 4-bromo-3-flourophenol, 3-fluoro-4-bromo phenol, 3-fluoro-4-bromo-phenol, 4-bromo-3-fluoro phenol, 4-bromo-5-fluoro-phenol, acmc-209a9m |
| PubChem CID | 2779153 |
| IUPAC Name | 4-bromo-3-fluorophenol |
| SMILES | C1=CC(=C(C=C1O)F)Br |
Chemical Identifiers
| 121219-03-2 | |
| 190.999 | |
| MRQYTJXVULSNIS-UHFFFAOYSA-N | |
| 2779153 | |
| C1=CC(=C(C=C1O)F)Br |
| C6H4BrFO | |
| MFCD00051907 | |
| 3-fluoro-4-bromophenol, 4-bromo-3-fluoro-phenol, phenol, 4-bromo-3-fluoro, pubchem1489, 4-bromo-3-flourophenol, 3-fluoro-4-bromo phenol, 3-fluoro-4-bromo-phenol, 4-bromo-3-fluoro phenol, 4-bromo-5-fluoro-phenol, acmc-209a9m | |
| 4-bromo-3-fluorophenol |
Specifications
| 121219-03-2 | |
| 96°C | |
| MFCD00051907 | |
| 3-fluoro-4-bromophenol, 4-bromo-3-fluoro-phenol, phenol, 4-bromo-3-fluoro, pubchem1489, 4-bromo-3-flourophenol, 3-fluoro-4-bromo phenol, 3-fluoro-4-bromo-phenol, 4-bromo-3-fluoro phenol, 4-bromo-5-fluoro-phenol, acmc-209a9m | |
| C1=CC(=C(C=C1O)F)Br | |
| 190.999 | |
| 191.00 | |
| Crystalline Powder |
| 73°C | |
| C6H4BrFO | |
| 5 g | |
| MRQYTJXVULSNIS-UHFFFAOYSA-N | |
| 4-bromo-3-fluorophenol | |
| 2779153 | |
| ≥98.0% (GC,T) | |
| 4-Bromo-3-fluorophenol |
Safety and Handling
TSCA : No