missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorophenol 98.0+%, TCI America™

Catalog No. B36885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36885G 5 g
B368825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36885G Supplier TCI America Supplier No. B36885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121219-03-2
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD00051907
InChI Key MRQYTJXVULSNIS-UHFFFAOYSA-N
Synonym 3-fluoro-4-bromophenol, 4-bromo-3-fluoro-phenol, phenol, 4-bromo-3-fluoro, pubchem1489, 4-bromo-3-flourophenol, 3-fluoro-4-bromo phenol, 3-fluoro-4-bromo-phenol, 4-bromo-3-fluoro phenol, 4-bromo-5-fluoro-phenol, acmc-209a9m
PubChem CID 2779153
IUPAC Name 4-bromo-3-fluorophenol
SMILES C1=CC(=C(C=C1O)F)Br

Specifications

Melting Point 73°C
Boiling Point 96°C
Quantity 5 g
Formula Weight 191.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-fluorophenol