missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorophenol, 98%

CAS: 121219-03-2 | C6H4BrFO | 190.999 g/mol

$114.74 - $370.71

Chemical Identifiers

CAS 121219-03-2
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD00051907
InChI Key MRQYTJXVULSNIS-UHFFFAOYSA-N
Synonym 3-fluoro-4-bromophenol, 4-bromo-3-fluoro-phenol, phenol, 4-bromo-3-fluoro, pubchem1489, 4-bromo-3-flourophenol, 3-fluoro-4-bromo phenol, 3-fluoro-4-bromo-phenol, 4-bromo-3-fluoro phenol, 4-bromo-5-fluoro-phenol, acmc-209a9m
PubChem CID 2779153
IUPAC Name 4-bromo-3-fluorophenol
SMILES C1=CC(=C(C=C1O)F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3248806
View Documents
Thermo Scientific Chemicals
H3248806
5 g
Each for $114.74
Only null left
 
AAH3248814
View Documents
Thermo Scientific Chemicals
H3248814
25 g
Each for $370.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121219-03-2
190.999
MRQYTJXVULSNIS-UHFFFAOYSA-N
2779153
C1=CC(=C(C=C1O)F)Br
C6H4BrFO
MFCD00051907
3-fluoro-4-bromophenol, 4-bromo-3-fluoro-phenol, phenol, 4-bromo-3-fluoro, pubchem1489, 4-bromo-3-flourophenol, 3-fluoro-4-bromo phenol, 3-fluoro-4-bromo-phenol, 4-bromo-3-fluoro phenol, 4-bromo-5-fluoro-phenol, acmc-209a9m
4-bromo-3-fluorophenol

Specifications

121219-03-2
C6H4BrFO
5 g
MRQYTJXVULSNIS-UHFFFAOYSA-N
4-bromo-3-fluorophenol
2779153
98%
71°C to 73°C
MFCD00051907
3-fluoro-4-bromophenol, 4-bromo-3-fluoro-phenol, phenol, 4-bromo-3-fluoro, pubchem1489, 4-bromo-3-flourophenol, 3-fluoro-4-bromo phenol, 3-fluoro-4-bromo-phenol, 4-bromo-3-fluoro phenol, 4-bromo-5-fluoro-phenol, acmc-209a9m
C1=CC(=C(C=C1O)F)Br
190.999
191
4-Bromo-3-fluorophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only