missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3-fluorobenzonitrile 98.0+%, TCI America™
$119.30 - $408.22
Chemical Identifiers
| CAS | 133059-44-6 |
|---|---|
| Molecular Formula | C7H3BrFN |
| Molecular Weight (g/mol) | 200.01 |
| MDL Number | MFCD07782069 |
| InChI Key | QBKXYSXQKRNVRQ-UHFFFAOYSA-N |
| Synonym | 3-fluoro-4-bromobenzonitrile, 4-cyano-2-fluorobromobenzene, 4-bromo-3-fluorobenzenecarbonitrile, benzonitrile, 4-bromo-3-fluoro, pubchem4707, intermediates-zcf02028, acmc-1c4z6, ksc493i5f, 4-bromo-3-fluoro-benzonitrile, 4-bromo-3-fluorobenzonitrile |
| PubChem CID | 15165443 |
| IUPAC Name | 4-bromo-3-fluorobenzonitrile |
| SMILES | C1=CC(=C(C=C1C#N)F)Br |
Chemical Identifiers
| 133059-44-6 | |
| 200.01 | |
| QBKXYSXQKRNVRQ-UHFFFAOYSA-N | |
| 15165443 | |
| C1=CC(=C(C=C1C#N)F)Br |
| C7H3BrFN | |
| MFCD07782069 | |
| 3-fluoro-4-bromobenzonitrile, 4-cyano-2-fluorobromobenzene, 4-bromo-3-fluorobenzenecarbonitrile, benzonitrile, 4-bromo-3-fluoro, pubchem4707, intermediates-zcf02028, acmc-1c4z6, ksc493i5f, 4-bromo-3-fluoro-benzonitrile, 4-bromo-3-fluorobenzonitrile | |
| 4-bromo-3-fluorobenzonitrile |
Specifications
| 133059-44-6 | |
| White-Yellow | |
| MFCD07782069 | |
| 3-fluoro-4-bromobenzonitrile, 4-cyano-2-fluorobromobenzene, 4-bromo-3-fluorobenzenecarbonitrile, benzonitrile, 4-bromo-3-fluoro, pubchem4707, intermediates-zcf02028, acmc-1c4z6, ksc493i5f, 4-bromo-3-fluoro-benzonitrile, 4-bromo-3-fluorobenzonitrile | |
| C1=CC(=C(C=C1C#N)F)Br | |
| 200.01 | |
| 200.01 | |
| Crystalline Powder |
| 94°C | |
| C7H3BrFN | |
| 5 g | |
| QBKXYSXQKRNVRQ-UHFFFAOYSA-N | |
| 4-bromo-3-fluorobenzonitrile | |
| 15165443 | |
| ≥98.0% (GC) | |
| 4-Bromo-3-fluorobenzonitrile |
Safety and Handling
TSCA : No