missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorobenzeneboronic acid, 95%

Catalog No. p-7047280
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL18514MD 250 mg
AAL1851403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL18514MD Supplier Thermo Scientific Chemicals Supplier No. L18514MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 374790-97-3 | C6H5BBrFO2 | 218.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 374790-97-3
Molecular Formula C6H5BBrFO2
Molecular Weight (g/mol) 218.82
MDL Number MFCD02093066
InChI Key ACLQPRPXJMWADE-UHFFFAOYSA-N
Synonym 4-bromo-3-fluorobenzeneboronic acid, 4-bromo-3-fluorophenyl boronic acid, boronic acid, 4-bromo-3-fluorophenyl, boronic acid, b-4-bromo-3-fluorophenyl, pubchem7779, 4-bromo-3-fluoro-phenyl boronic acid, acmc-209it9, 4-bromo-3-fluorobenzeneboronicacid
PubChem CID 2773335
IUPAC Name (4-bromo-3-fluorophenyl)boronic acid
SMILES OB(O)C1=CC=C(Br)C(F)=C1

Specifications

Melting Point >300°C
Quantity 250 mg
Formula Weight 218.82
Percent Purity 95%
Chemical Name or Material 4-Bromo-3-fluorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only