missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorobenzaldehyde 98.0+%, TCI America™

Catalog No. p-7123467
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43195G 5 g
B431925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43195G Supplier TCI America Supplier No. B43195G
Only null left

Chemical Identifiers

CAS 133059-43-5
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD03095000
InChI Key SWHUROFMIMHWKS-UHFFFAOYSA-N
Synonym 3-fluoro-4-bromobenzaldehyde, 3-fluoro-4-bromo benzaldehyde, 4-bromo-3-fluoro-benzaldehyde, benzaldehyde, 4-bromo-3-fluoro, 4-bromo-3-fluoro benzaldehyde, pubchem4033, acmc-209bqp, intermediates-zcf02027, ksc174m4l, 3-fluoro-4-bromo-benzaldehyde
PubChem CID 2783411
IUPAC Name 4-bromo-3-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)F)Br

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 5 g
Formula Weight 203.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-fluorobenzaldehyde