missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluoroanisole 98.0+%, TCI America™

Catalog No. p-7123137
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37435G 5 g
B374325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37435G Supplier TCI America Supplier No. B37435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 458-50-4
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.026
MDL Number MFCD01310983
InChI Key XANVIFOBBVAKCY-UHFFFAOYSA-N
Synonym 4-bromo-3-fluoroanisole, 3-fluoro-4-bromo anisole, 3-fluoro-4-bromoanisol, 3-fluoro-4-bromo anisol, 1-bromo-2-fluoro-4-methoxy-benzene, benzene, 1-bromo-2-fluoro-4-methoxy, pharmabridge p-1018, rarechem al mz 0889, zlchem 365, pubchem1970
PubChem CID 3718444
IUPAC Name 1-bromo-2-fluoro-4-methoxybenzene
SMILES COC1=CC(=C(C=C1)Br)F

Specifications

Color Yellow
Boiling Point 216°C
Quantity 5 g
Formula Weight 205.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-3-fluoroanisole