missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3-fluoroaniline 98.0+%, TCI America™
$44.73 - $121.16
Chemical Identifiers
| CAS | 656-65-5 |
|---|---|
| Molecular Formula | C6H5BrFN |
| Molecular Weight (g/mol) | 190.015 |
| MDL Number | MFCD00672933 |
| InChI Key | YTMVYYAKOPIJCZ-UHFFFAOYSA-N |
| Synonym | 4-bromo-3-fluorophenylamine, 3-fluoro-4-bromoaniline, 4-bromo-3-fluoro aniline, benzenamine, 4-bromo-3-fluoro, 4-brom-3-fluoranilin, zlchem 544, pubchem1526, acmc-1badx, 4-bromo-3-fluro-aniline, 4-bromo-3-fluoro-aniline |
| PubChem CID | 821848 |
| IUPAC Name | 4-bromo-3-fluoroaniline |
| SMILES | C1=CC(=C(C=C1N)F)Br |
Chemical Identifiers
| 656-65-5 | |
| 190.015 | |
| YTMVYYAKOPIJCZ-UHFFFAOYSA-N | |
| 821848 | |
| C1=CC(=C(C=C1N)F)Br |
| C6H5BrFN | |
| MFCD00672933 | |
| 4-bromo-3-fluorophenylamine, 3-fluoro-4-bromoaniline, 4-bromo-3-fluoro aniline, benzenamine, 4-bromo-3-fluoro, 4-brom-3-fluoranilin, zlchem 544, pubchem1526, acmc-1badx, 4-bromo-3-fluro-aniline, 4-bromo-3-fluoro-aniline | |
| 4-bromo-3-fluoroaniline |
Specifications
| 656-65-5 | |
| C6H5BrFN | |
| 1 g | |
| YTMVYYAKOPIJCZ-UHFFFAOYSA-N | |
| 4-bromo-3-fluoroaniline | |
| 821848 | |
| ≥98.0% (GC) | |
| 4-Bromo-3-fluoroaniline |
| 72°C | |
| MFCD00672933 | |
| 4-bromo-3-fluorophenylamine, 3-fluoro-4-bromoaniline, 4-bromo-3-fluoro aniline, benzenamine, 4-bromo-3-fluoro, 4-brom-3-fluoranilin, zlchem 544, pubchem1526, acmc-1badx, 4-bromo-3-fluro-aniline, 4-bromo-3-fluoro-aniline | |
| C1=CC(=C(C=C1N)F)Br | |
| 190.015 | |
| 190.02 | |
| Crystalline Powder |
Safety and Handling
TSCA : No