missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3-fluoroaniline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 72°C |
| Quantity | 1 g |
| Formula Weight | 190.02 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Bromo-3-fluoroaniline |
Safety and Handling
| TSCA | No |