missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-3-chloropropiophenone, 94%

Catalog No. AAA1398809
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1398809 10 g
AAA1398818 50 g
AAA1398830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1398809 Supplier Thermo Scientific Chemicals Supplier No. A1398809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 31736-73-9 | C9H8BrClO | 247.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31736-73-9
Molecular Formula C9H8BrClO
Molecular Weight (g/mol) 247.52
MDL Number MFCD00018979
InChI Key SYWQQEVHZPPLNQ-UHFFFAOYSA-N
Synonym 4'-bromo-3-chloropropiophenone, 1-4-bromophenyl-3-chloropropan-1-one, 4-bromo-3-chloropropiophenone, 1-propanone, 1-4-bromophenyl-3-chloro, pubchem13267, ksc567m6b, 3-chloro-4'-bromopropiophenone, 4'-bromo-beta-chloropropiophenone, 4-3-chloropropanoyl bromobenzene, 4\\'-bromo-3-chloropropiophenone
PubChem CID 95790
IUPAC Name 1-(4-bromophenyl)-3-chloropropan-1-one
SMILES ClCCC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 57°C to 62°C
Quantity 10 g
Beilstein 1944367
Formula Weight 247.53
Percent Purity 94%
Chemical Name or Material 4'-Bromo-3-chloropropiophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 250-784-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only