accessibility menu, dialog, popup

UserName

ClOBr

CAS RN 31736-73-9

IUPAC Name: 1-(4-bromophenyl)-3-chloropropan-1-one
Synonyms: 1-(4-bromophenyl)-3-chloropropan-1-one
Molecular Weight (g/mol): 247.52
Molecular Formula: C9H8BrClO
InChi Key: SYWQQEVHZPPLNQ-UHFFFAOYSA-N
SMILES: ClCCC(=O)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)