missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3-chlorobenzoic Acid 98.0+%, TCI America™
$89.48 - $356.03
Chemical Identifiers
| CAS | 25118-59-6 |
|---|---|
| Molecular Formula | C7H4BrClO2 |
| Molecular Weight (g/mol) | 235.46 |
| MDL Number | MFCD08276864 |
| InChI Key | PSKJIHDVFDVNBU-UHFFFAOYSA-N |
| Synonym | benzoic acid, 4-bromo-3-chloro, acmc-1cbza, intermediates-zcf02677, ksc201k8d, 3-chloro-4-bromobenzoic acid, 4-bromo-3-chloro-benzoic acid, 3-chloro-4-bromor-benzoic acid, benzoicacid, 4-bromo-3-chloro, 4-bromanyl-3-chloranyl-benzoic acid, 4-bromo-3-chlorobenzoic acid |
| PubChem CID | 12594302 |
| IUPAC Name | 4-bromo-3-chlorobenzoic acid |
| SMILES | OC(=O)C1=CC(Cl)=C(Br)C=C1 |
Chemical Identifiers
| 25118-59-6 | |
| 235.46 | |
| PSKJIHDVFDVNBU-UHFFFAOYSA-N | |
| 12594302 | |
| OC(=O)C1=CC(Cl)=C(Br)C=C1 |
| C7H4BrClO2 | |
| MFCD08276864 | |
| benzoic acid, 4-bromo-3-chloro, acmc-1cbza, intermediates-zcf02677, ksc201k8d, 3-chloro-4-bromobenzoic acid, 4-bromo-3-chloro-benzoic acid, 3-chloro-4-bromor-benzoic acid, benzoicacid, 4-bromo-3-chloro, 4-bromanyl-3-chloranyl-benzoic acid, 4-bromo-3-chlorobenzoic acid | |
| 4-bromo-3-chlorobenzoic acid |
Specifications
| 25118-59-6 | |
| White-Yellow | |
| MFCD08276864 | |
| benzoic acid, 4-bromo-3-chloro, acmc-1cbza, intermediates-zcf02677, ksc201k8d, 3-chloro-4-bromobenzoic acid, 4-bromo-3-chloro-benzoic acid, 3-chloro-4-bromor-benzoic acid, benzoicacid, 4-bromo-3-chloro, 4-bromanyl-3-chloranyl-benzoic acid, 4-bromo-3-chlorobenzoic acid | |
| OC(=O)C1=CC(Cl)=C(Br)C=C1 | |
| 235.46 | |
| 235.46 | |
| Crystalline Powder |
| 222°C | |
| C7H4BrClO2 | |
| 5 g | |
| PSKJIHDVFDVNBU-UHFFFAOYSA-N | |
| 4-bromo-3-chlorobenzoic acid | |
| 12594302 | |
| ≥98.0% (GC,T) | |
| 4-Bromo-3-chlorobenzoic Acid |
Safety and Handling
TSCA : No