missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-chlorobenzoic Acid 98.0+%, TCI America™

Catalog No. B37915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37915G 5 g
B379125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37915G Supplier TCI America Supplier No. B37915G

Chemical Identifiers

CAS 25118-59-6
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.46
MDL Number MFCD08276864
InChI Key PSKJIHDVFDVNBU-UHFFFAOYSA-N
Synonym benzoic acid, 4-bromo-3-chloro, acmc-1cbza, intermediates-zcf02677, ksc201k8d, 3-chloro-4-bromobenzoic acid, 4-bromo-3-chloro-benzoic acid, 3-chloro-4-bromor-benzoic acid, benzoicacid, 4-bromo-3-chloro, 4-bromanyl-3-chloranyl-benzoic acid, 4-bromo-3-chlorobenzoic acid
PubChem CID 12594302
IUPAC Name 4-bromo-3-chlorobenzoic acid
SMILES OC(=O)C1=CC(Cl)=C(Br)C=C1

Specifications

Melting Point 222°C
Color White-Yellow
Quantity 5 g
Formula Weight 235.46
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-chlorobenzoic Acid