missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-chlorobenzoic acid, 97%

Catalog No. AAH2725214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2725214 25 g
AAH2725206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2725214 Supplier Thermo Scientific Chemicals Supplier No. H2725214

CAS: 25118-59-6 | C7H4BrClO2 | 235.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25118-59-6
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.46
MDL Number MFCD08276864
InChI Key PSKJIHDVFDVNBU-UHFFFAOYSA-N
Synonym benzoic acid, 4-bromo-3-chloro, acmc-1cbza, intermediates-zcf02677, ksc201k8d, 3-chloro-4-bromobenzoic acid, 4-bromo-3-chloro-benzoic acid, 3-chloro-4-bromor-benzoic acid, benzoicacid, 4-bromo-3-chloro, 4-bromanyl-3-chloranyl-benzoic acid, 4-bromo-3-chlorobenzoic acid
PubChem CID 12594302
IUPAC Name 4-bromo-3-chlorobenzoic acid
SMILES OC(=O)C1=CC(Cl)=C(Br)C=C1

Specifications

Melting Point 220°C to 225°C
Odor Odorless
Quantity 25 g
Formula Weight 235.47
Percent Purity 97%
Chemical Name or Material 4-Bromo-3-chlorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only