missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-chloroaniline 98.0+%, TCI America™

Catalog No. p-7123113
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37095G 5 g
B370925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37095G Supplier TCI America Supplier No. B37095G
Only null left

Chemical Identifiers

CAS 21402-26-6
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD00041311
InChI Key QLYHPNUFNZJXOQ-UHFFFAOYSA-N
Synonym benzenamine, 4-bromo-3-chloro, 3-chloro-4-bromoaniline, unii-2gtm4sw4oh, 2gtm4sw4oh, 4-bromo-3-chloro-aniline, 4-bromo-3-chlorophenyl amine, pubchem3579, zlchem 1051, 4-bromo-3-chloro aniline, 4-bromo-3-chlorophenylamine
PubChem CID 88888
IUPAC Name 4-bromo-3-chloroaniline
SMILES C1=CC(=C(C=C1N)Cl)Br

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 5 g
Formula Weight 206.47
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-chloroaniline