missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-chloroaniline, 96%

Catalog No. p-7043879
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0326809 10 g
AAL0326818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0326809 Supplier Thermo Scientific Chemicals Supplier No. L0326809
Only null left

CAS: 21402-26-6 | C6H5BrClN | 206.467 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21402-26-6
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD00041311
InChI Key QLYHPNUFNZJXOQ-UHFFFAOYSA-N
Synonym benzenamine, 4-bromo-3-chloro, 3-chloro-4-bromoaniline, unii-2gtm4sw4oh, 2gtm4sw4oh, 4-bromo-3-chloro-aniline, 4-bromo-3-chlorophenyl amine, pubchem3579, zlchem 1051, 4-bromo-3-chloro aniline, 4-bromo-3-chlorophenylamine
PubChem CID 88888
IUPAC Name 4-bromo-3-chloroaniline
SMILES C1=CC(=C(C=C1N)Cl)Br

Specifications

Melting Point 61°C to 66°C
Odor Characteristic
Quantity 10 g
UN Number UN2811
Beilstein 1210160
Formula Weight 206.47
Percent Purity 96%
Chemical Name or Material 4-Bromo-3-chloroaniline
Hazard Category H302+H332-H311-H373
Hazard Statement GHS H Statement
H311-H373-H302-H332
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 244-370-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only