missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-3'-chloroacetanilide 98.0+%, TCI America™

Catalog No. B31415G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31415G 5 g
B314125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31415G Supplier TCI America Supplier No. B31415G

Chemical Identifiers

CAS 22459-81-0
Molecular Formula C8H7BrClNO
Molecular Weight (g/mol) 248.504
MDL Number MFCD00040848
InChI Key BVADKOYGDKTWMU-UHFFFAOYSA-N
Synonym 4'-bromo-3'-chloroacetanilide, n-4-bromo-3-chlorophenyl acetamide, n-acetyl 4-bromo-3-chloroaniline, 4-bromo-3-chloroacetanilide, n1-4-bromo-3-chlorophenyl acetamide, acetamide,n-4-bromo-3-chlorophenyl, acmc-209fwl, maybridge1_008946, n-acetyl4-bromo-3-chloroaniline
PubChem CID 2746295
IUPAC Name N-(4-bromo-3-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC(=C(C=C1)Br)Cl

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 5 g
Formula Weight 248.50
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromo-3′-chloroacetanilide