accessibility menu, dialog, popup

UserName

CH3ONHBrCl

CAS RN 22459-81-0

IUPAC Name: N-(4-bromo-3-chlorophenyl)acetamide
Synonyms: N-(4-bromo-3-chlorophenyl)acetamide
Molecular Weight (g/mol): 248.5
Molecular Formula: C8H7BrClNO
InChi Key: BVADKOYGDKTWMU-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(Br)C(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results