missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethylisoxazole, 97%, Thermo Scientific™

Chemical Identifiers

CAS 10558-25-5
Molecular Formula C5H6BrNO
Molecular Weight (g/mol) 176.013
MDL Number MFCD00068187
InChI Key GYHZPSUAMYIFQD-UHFFFAOYSA-N
Synonym 4-bromo-3,5-dimethylisoxazole, 3,5-dimethyl-4-bromoisoxazole, isoxazole, 4-bromo-3,5-dimethyl, 4-bromo-dimethylisoxazole, buttpark 33\06-98, 4-bromo-3,5-dimethylisoxazole, tech., pubchem8720, acmc-1bpqs, timtec-bb sbb003784, ksc182i2n
PubChem CID 318421
IUPAC Name 4-bromo-3,5-dimethyl-1,2-oxazole
SMILES CC1=C(C(=NO1)C)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC00610DA
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

10558-25-5
176.013
GYHZPSUAMYIFQD-UHFFFAOYSA-N
318421
CC1=C(C(=NO1)C)Br
C5H6BrNO
MFCD00068187
4-bromo-3,5-dimethylisoxazole, 3,5-dimethyl-4-bromoisoxazole, isoxazole, 4-bromo-3,5-dimethyl, 4-bromo-dimethylisoxazole, buttpark 33\06-98, 4-bromo-3,5-dimethylisoxazole, tech., pubchem8720, acmc-1bpqs, timtec-bb sbb003784, ksc182i2n
4-bromo-3,5-dimethyl-1,2-oxazole

Specifications

10558-25-5
100.0
97%
C5H6BrNO
4-bromo-3,5-dimethylisoxazole, 3,5-dimethyl-4-bromoisoxazole, isoxazole, 4-bromo-3,5-dimethyl, 4-bromo-dimethylisoxazole, buttpark 33\06-98, 4-bromo-3,5-dimethylisoxazole, tech., pubchem8720, acmc-1bpqs, timtec-bb sbb003784, ksc182i2n
CC1=C(C(=NO1)C)Br
176.013
176.01
4-Bromo-3, 5-dimethylisoxazole
96.5
1.484g/mL
Amber glass bottle
MFCD00068187
GYHZPSUAMYIFQD-UHFFFAOYSA-N
4-bromo-3,5-dimethyl-1,2-oxazole
318421
97%
Videos
SDS
Documents

Documents