missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethoxybenzaldehyde 98.0+%, TCI America™

Catalog No. p-7119857
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29525G 5 g
B295225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29525G Supplier TCI America Supplier No. B29525G
Only null left

Chemical Identifiers

CAS 31558-40-4
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.072
MDL Number MFCD00040799
InChI Key UGBJRYUNSXFPOX-UHFFFAOYSA-N
PubChem CID 3015579
IUPAC Name 4-bromo-3,5-dimethoxybenzaldehyde
SMILES COC1=CC(=CC(=C1Br)OC)C=O

Specifications

Melting Point 115°C
Quantity 5 g
Formula Weight 245.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3,5-dimethoxybenzaldehyde