missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3,3,4,4-tetrafluoro-1-butene 98.0+%, TCI America™
Supplier: TCI America B32225G
Specifications
| 4-Bromo-3,3,4,4-tetrafluoro-1-butene | |
| Yellow | |
| C4H3BrF4 | |
| 5 g | |
| GVCWGFZDSIWLMO-UHFFFAOYSA-N | |
| 4-bromo-3,3,4,4-tetrafluorobut-1-ene | |
| 87721 | |
| ≥98.0% (GC) |
| 18599-22-9 | |
| 60°C | |
| MFCD00042189 | |
| 4-bromo-3,3,4,4-tetrafluoro-1-butene, 1-butene, 4-bromo-3,3,4,4-tetrafluoro, unii-t08c3oeq6e, t08c3oeq6e, 4-bromo-3,3,4,4-tetrafluorobutene-1, 1-butene,4-bromo-3,3,4,4-tetrafluoro, acmc-209enf, gvcwgfzdsiwlmo-uhfffaoysa, 4-bromo-3,3,4,4-tetrafluorobutene, 1-bromo-1,1,2,2-tetrafluoro-3-butene | |
| FC(F)(Br)C(F)(F)C=C | |
| 206.97 | |
| 206.97 | |
| Liquid |
Chemical Identifiers
| 18599-22-9 | |
| 206.97 | |
| GVCWGFZDSIWLMO-UHFFFAOYSA-N | |
| 87721 | |
| FC(F)(Br)C(F)(F)C=C |
| C4H3BrF4 | |
| MFCD00042189 | |
| 4-bromo-3,3,4,4-tetrafluoro-1-butene, 1-butene, 4-bromo-3,3,4,4-tetrafluoro, unii-t08c3oeq6e, t08c3oeq6e, 4-bromo-3,3,4,4-tetrafluorobutene-1, 1-butene,4-bromo-3,3,4,4-tetrafluoro, acmc-209enf, gvcwgfzdsiwlmo-uhfffaoysa, 4-bromo-3,3,4,4-tetrafluorobutene, 1-bromo-1,1,2,2-tetrafluoro-3-butene | |
| 4-bromo-3,3,4,4-tetrafluorobut-1-ene |
Safety and Handling
TSCA : Yes