missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-(trifluoromethyl)phenol 97.0+%, TCI America™
$121.28 - $481.30
Chemical Identifiers
| CAS | 50824-04-9 |
|---|---|
| Molecular Formula | C7H4BrF3O |
| Molecular Weight (g/mol) | 241.007 |
| MDL Number | MFCD05668981 |
| InChI Key | PDPGERGWEOJVDC-UHFFFAOYSA-N |
| Synonym | 4-bromo-2-trifluoromethyl phenol, 4-bromo-2-trifluoromethyl benzenol, 5-bromo-2-hydroxybenzotrifluoride, o-cresol, 4-bromo-alpha,alpha,alpha-trifluoro, 4-bromo-2-trifluoromethyl-phenol, phenol, 4-bromo-2-trifluoromethyl, pubchem18454, acmc-209kqd, bromotrifluoromethylbenzenol, ksc490k9d |
| PubChem CID | 142784 |
| IUPAC Name | 4-bromo-2-(trifluoromethyl)phenol |
| SMILES | C1=CC(=C(C=C1Br)C(F)(F)F)O |
Chemical Identifiers
| 50824-04-9 | |
| 241.007 | |
| PDPGERGWEOJVDC-UHFFFAOYSA-N | |
| 142784 | |
| C1=CC(=C(C=C1Br)C(F)(F)F)O |
| C7H4BrF3O | |
| MFCD05668981 | |
| 4-bromo-2-trifluoromethyl phenol, 4-bromo-2-trifluoromethyl benzenol, 5-bromo-2-hydroxybenzotrifluoride, o-cresol, 4-bromo-alpha,alpha,alpha-trifluoro, 4-bromo-2-trifluoromethyl-phenol, phenol, 4-bromo-2-trifluoromethyl, pubchem18454, acmc-209kqd, bromotrifluoromethylbenzenol, ksc490k9d | |
| 4-bromo-2-(trifluoromethyl)phenol |
Specifications
| 50824-04-9 | |
| White-Yellow | |
| MFCD05668981 | |
| 4-bromo-2-trifluoromethyl phenol, 4-bromo-2-trifluoromethyl benzenol, 5-bromo-2-hydroxybenzotrifluoride, o-cresol, 4-bromo-alpha,alpha,alpha-trifluoro, 4-bromo-2-trifluoromethyl-phenol, phenol, 4-bromo-2-trifluoromethyl, pubchem18454, acmc-209kqd, bromotrifluoromethylbenzenol, ksc490k9d | |
| C1=CC(=C(C=C1Br)C(F)(F)F)O | |
| 241.007 | |
| 241.01 | |
| Crystalline Powder |
| 85°C | |
| C7H4BrF3O | |
| 5 g | |
| PDPGERGWEOJVDC-UHFFFAOYSA-N | |
| 4-bromo-2-(trifluoromethyl)phenol | |
| 142784 | |
| ≥97.0% (GC) | |
| 4-Bromo-2-(trifluoromethyl)phenol |
Safety and Handling
TSCA : No