missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethyl)phenol 97.0+%, TCI America™

Catalog No. B44935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44935G 5 g
B449325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44935G Supplier TCI America Supplier No. B44935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50824-04-9
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
MDL Number MFCD05668981
InChI Key PDPGERGWEOJVDC-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethyl phenol, 4-bromo-2-trifluoromethyl benzenol, 5-bromo-2-hydroxybenzotrifluoride, o-cresol, 4-bromo-alpha,alpha,alpha-trifluoro, 4-bromo-2-trifluoromethyl-phenol, phenol, 4-bromo-2-trifluoromethyl, pubchem18454, acmc-209kqd, bromotrifluoromethylbenzenol, ksc490k9d
PubChem CID 142784
IUPAC Name 4-bromo-2-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1Br)C(F)(F)F)O

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 5 g
Formula Weight 241.01
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)phenol