missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethyl)phenol, 99%

Catalog No. AAH3178703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3178703 1 g
AAH3178706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3178703 Supplier Thermo Scientific Chemicals Supplier No. H3178703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50824-04-9 | C7H4BrF3O | 241.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50824-04-9
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
MDL Number MFCD05668981
InChI Key PDPGERGWEOJVDC-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethyl phenol, 4-bromo-2-trifluoromethyl benzenol, 5-bromo-2-hydroxybenzotrifluoride, o-cresol, 4-bromo-alpha,alpha,alpha-trifluoro, 4-bromo-2-trifluoromethyl-phenol, phenol, 4-bromo-2-trifluoromethyl, pubchem18454, acmc-209kqd, bromotrifluoromethylbenzenol, ksc490k9d
PubChem CID 142784
IUPAC Name 4-bromo-2-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1Br)C(F)(F)F)O

Specifications

Melting Point 82°C to 86°C
Quantity 1 g
Formula Weight 241.01
Percent Purity 99%
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)phenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only