missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™
$105.89 - $419.69
Chemical Identifiers
| CAS | 320-31-0 |
|---|---|
| Molecular Formula | C8H4BrF3O2 |
| Molecular Weight (g/mol) | 269.02 |
| MDL Number | MFCD07787468 |
| InChI Key | JXHWAPDBDXPBEQ-UHFFFAOYSA-N |
| Synonym | 4-bromo-2-trifluoromethyl benzoic acid, benzoic acid, 4-bromo-2-trifluoromethyl, 2-trifluoromethyl-4-bromobenzoic acid, pubchem4738, 4-bromo-2-trifluoromethyl benzoicacid, acmc-209hq4, ksc495g7n, 5-bromo-2-carboxybenzotrifluoride |
| PubChem CID | 25067392 |
| IUPAC Name | 4-bromo-2-(trifluoromethyl)benzoic acid |
| SMILES | OC(=O)C1=C(C=C(Br)C=C1)C(F)(F)F |
Chemical Identifiers
| 320-31-0 | |
| 269.02 | |
| JXHWAPDBDXPBEQ-UHFFFAOYSA-N | |
| 25067392 | |
| OC(=O)C1=C(C=C(Br)C=C1)C(F)(F)F |
| C8H4BrF3O2 | |
| MFCD07787468 | |
| 4-bromo-2-trifluoromethyl benzoic acid, benzoic acid, 4-bromo-2-trifluoromethyl, 2-trifluoromethyl-4-bromobenzoic acid, pubchem4738, 4-bromo-2-trifluoromethyl benzoicacid, acmc-209hq4, ksc495g7n, 5-bromo-2-carboxybenzotrifluoride | |
| 4-bromo-2-(trifluoromethyl)benzoic acid |
Specifications
| 320-31-0 | |
| White | |
| MFCD07787468 | |
| 4-bromo-2-trifluoromethyl benzoic acid, benzoic acid, 4-bromo-2-trifluoromethyl, 2-trifluoromethyl-4-bromobenzoic acid, pubchem4738, 4-bromo-2-trifluoromethyl benzoicacid, acmc-209hq4, ksc495g7n, 5-bromo-2-carboxybenzotrifluoride | |
| OC(=O)C1=C(C=C(Br)C=C1)C(F)(F)F | |
| 269.02 | |
| 269.02 | |
| Crystalline Powder |
| 123°C | |
| C8H4BrF3O2 | |
| 1 g | |
| JXHWAPDBDXPBEQ-UHFFFAOYSA-N | |
| 4-bromo-2-(trifluoromethyl)benzoic acid | |
| 25067392 | |
| ≥98.0% (GC,T) | |
| 4-Bromo-2-(trifluoromethyl)benzoic Acid |
Safety and Handling
TSCA : No