missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™

Catalog No. B48431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48431G 1 g
B48435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48431G Supplier TCI America Supplier No. B48431G

Chemical Identifiers

CAS 320-31-0
Molecular Formula C8H4BrF3O2
Molecular Weight (g/mol) 269.02
MDL Number MFCD07787468
InChI Key JXHWAPDBDXPBEQ-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethyl benzoic acid, benzoic acid, 4-bromo-2-trifluoromethyl, 2-trifluoromethyl-4-bromobenzoic acid, pubchem4738, 4-bromo-2-trifluoromethyl benzoicacid, acmc-209hq4, ksc495g7n, 5-bromo-2-carboxybenzotrifluoride
PubChem CID 25067392
IUPAC Name 4-bromo-2-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=C(C=C(Br)C=C1)C(F)(F)F

Specifications

Melting Point 123°C
Color White
Quantity 1 g
Formula Weight 269.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)benzoic Acid