missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethoxy)phenol, 98%, Thermo Scientific Chemicals

Catalog No. AAH3172606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3172606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3172606 Supplier Thermo Scientific Chemicals Supplier No. H3172606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 690264-39-2 | Molecular Formula: C7H4BrF3O2 | Molecular Weight: 257.01 g/mol

4-Bromo-2-(trifluoromethoxy)phenol are the basic building blocks numerous pharmaceutical drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-(trifluoromethoxy)phenol are the basic building blocks numerous pharmaceutical drugs.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 690264-39-2
Molecular Formula C7H4BrF3O2
Molecular Weight (g/mol) 257.01
MDL Number MFCD09037798
InChI Key GYXNTPVMXGBRQW-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethoxy phenol, 4-bromo-2-trifluoromethoxy-phenol, 4-bromanyl-2-trifluoromethyloxy phenol, phenol, 4-bromo-2-trifluoromethoxy
PubChem CID 11780151
IUPAC Name 4-bromo-2-(trifluoromethoxy)phenol
SMILES OC1=C(OC(F)(F)F)C=C(Br)C=C1

Specifications

Melting Point 51°C to 55°C
Quantity 5 g
Formula Weight 257.01
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-(trifluoromethoxy)phenol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only