missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethoxy)anisole, 98%

Catalog No. AAH3264503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3264503 1 g
AAH3264506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3264503 Supplier Thermo Scientific Chemicals Supplier No. H3264503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 853771-88-7 | C8H6BrF3O2 | 271.03 g/mol

4-Bromo-2-(trifluoromethoxy)anisole employed in biological evaluation of some analogs of the antitumor agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-(trifluoromethoxy)anisole employed in biological evaluation of some analogs of the antitumor agents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 853771-88-7
Molecular Formula C8H6BrF3O2
Molecular Weight (g/mol) 271.03
MDL Number MFCD09907950
InChI Key QVGIROOBNUFIKC-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethoxy anisole, 4-bromo-1-methoxy-2-trifluoromethoxy benzene, benzene,4-bromo-1-methoxy-2-trifluoromethoxy
PubChem CID 45790939
IUPAC Name 4-bromo-1-methoxy-2-(trifluoromethoxy)benzene
SMILES COC1=C(OC(F)(F)F)C=C(Br)C=C1

Specifications

Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 271.04
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-(trifluoromethoxy)anisole
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only