missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-tert-butylphenol 95.0+%, TCI America™

Catalog No. B43155G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43155G 5 g
B43151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43155G Supplier TCI America Supplier No. B43155G

Chemical Identifiers

CAS 10323-39-4
Molecular Formula C10H13BrO
Molecular Weight (g/mol) 229.12
MDL Number MFCD00192645
InChI Key IKMJSWBFODAWTC-UHFFFAOYSA-N
PubChem CID 97068
IUPAC Name 4-bromo-2-tert-butylphenol
SMILES CC(C)(C)C1=CC(Br)=CC=C1O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 229.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-tert-butylphenol
TSCA No
Recommended Storage Refrigerator