missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitrotoluene 98.0+%, TCI America™

Catalog No. p-7119300
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18435G 5 g
B184325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18435G Supplier TCI America Supplier No. B18435G

Chemical Identifiers

CAS 60956-26-5
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00041243
InChI Key KZNXALJXBRSMFL-UHFFFAOYSA-N
Synonym 4-bromo-2-nitrotoluene, 2-nitro-4-bromotoluene, 4-bromo-6-nitrotoluene, bromo 4--2-nitrotoluene, 5-bromo-2-methylnitrobenzene, benzene, 4-bromo-1-methyl-2-nitro, 4-bromo-1-methyl-2-nitro-benzene, 4-brom-2-nitrotoluol, pubchem10403, maybridge1_002072
PubChem CID 123546
IUPAC Name 4-bromo-1-methyl-2-nitrobenzene
SMILES CC1=C(C=C(C=C1)Br)[N+](=O)[O-]

Specifications

Melting Point 46°C
Boiling Point 256°C
Quantity 5 g
Formula Weight 216.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-nitrotoluene