missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7123180
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38105G 5 g
B381025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38105G Supplier TCI America Supplier No. B38105G
Only null left

Chemical Identifiers

CAS 99277-71-1
Molecular Formula C7H3BrNO4
Molecular Weight (g/mol) 245.01
MDL Number MFCD01013599
InChI Key ZIRHHEZLJGORGU-UHFFFAOYSA-M
Synonym 4-bromo-2-nitrobenzoic acid, 4-bromo-2-nitro-benzoic acid, 2-nitro-4-bromobenzoic acid, 5-bromo-2-carboxynitrobenzene, benzoic acid, 4-bromo-2-nitro, 4-bromo-2-nitrobenzoicacid, pubchem2055, 4-bromo-2-nitrobenzoic, acmc-209sc4, bromonitrobenzoicacid 42-
PubChem CID 3774467
IUPAC Name 4-bromo-2-nitrobenzoate
SMILES [O-]C(=O)C1=CC=C(Br)C=C1[N+]([O-])=O

Specifications

Melting Point 165°C
Quantity 5 g
Formula Weight 246.02
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-nitrobenzoic Acid