missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylpyridine, 97%

CAS: 22282-99-1 | C6H6BrN | 172.03 g/mol

Supplier:  Thermo Scientific Chemicals H2788403

 View more versions of this product

Catalog No. AAH2788403


Only null left
Explore available promotions

Description

Description

4-Bromo-2-methylpyridine is used as a starting material in the preparation of crown-ester-bipyridines and viologens through sodium or nickel reductive coupling, side chain oxidation and esterification. Further, it serves as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals and in dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-methylpyridine is used as a starting material in the preparation of crown-ester-bipyridines and viologens through sodium or nickel reductive coupling, side chain oxidation and esterification. Further, it serves as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals and in dyes.

Solubility
Miscible with dichloromethane. Immiscible with water.

Notes
Light and moisture sensitive. Incompatible with strong oxidizing agents, reducing agents, strong acids and bases. Store in a cool place.
Specifications

Specifications

4-Bromo-2-methylpyridine
1.45
79°C (174°F)
C6H6BrN
MFCD03788196
4-bromo-2-picoline, 2-methyl-4-bromopyridine, 4-bromo-2-methyl-pyridine, pyridine, 4-bromo-2-methyl, 4-bromopicoline, zlchem 422, pubchem4035, acmc-209fui, 4-bromo-methyl-pyridine
JFBMFWHEXBLFCR-UHFFFAOYSA-N
4-bromo-2-methylpyridine
2762835
97%
22282-99-1
76°C (14 mmHg)
Characteristic
1.556
1 g
Miscible with dichloromethane. Immiscible with water.
CC1=CC(Br)=CC=N1
172.03
172.03

Chemical Identifiers

22282-99-1
172.03
JFBMFWHEXBLFCR-UHFFFAOYSA-N
2762835
CC1=CC(Br)=CC=N1
C6H6BrN
MFCD03788196
4-bromo-2-picoline, 2-methyl-4-bromopyridine, 4-bromo-2-methyl-pyridine, pyridine, 4-bromo-2-methyl, 4-bromopicoline, zlchem 422, pubchem4035, acmc-209fui, 4-bromo-methyl-pyridine
4-bromo-2-methylpyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H227-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only