missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylpyridine, 97%

Catalog No. p-7030315
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27884MD 250 mg
AAH2788403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27884MD Supplier Thermo Scientific Chemicals Supplier No. H27884MD
Only null left

CAS: 22282-99-1 | C6H6BrN | 172.03 g/mol

4-Bromo-2-methylpyridine is used as a starting material in the preparation of crown-ester-bipyridines and viologens through sodium or nickel reductive coupling, side chain oxidation and esterification. Further, it serves as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals and in dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-methylpyridine is used as a starting material in the preparation of crown-ester-bipyridines and viologens through sodium or nickel reductive coupling, side chain oxidation and esterification. Further, it serves as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals and in dyes.

Solubility
Miscible with dichloromethane. Immiscible with water.

Notes
Light and moisture sensitive. Incompatible with strong oxidizing agents, reducing agents, strong acids and bases. Store in a cool place.

Chemical Identifiers

CAS 22282-99-1
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD03788196
InChI Key JFBMFWHEXBLFCR-UHFFFAOYSA-N
Synonym 4-bromo-2-picoline, 2-methyl-4-bromopyridine, 4-bromo-2-methyl-pyridine, pyridine, 4-bromo-2-methyl, 4-bromopicoline, zlchem 422, pubchem4035, acmc-209fui, 4-bromo-methyl-pyridine
PubChem CID 2762835
IUPAC Name 4-bromo-2-methylpyridine
SMILES CC1=CC(Br)=CC=N1

Specifications

Density 1.45
Boiling Point 76°C (14 mmHg)
Flash Point 79°C (174°F)
Odor Characteristic
Refractive Index 1.556
Quantity 250 mg
Solubility Information Miscible with dichloromethane. Immiscible with water.
Formula Weight 172.03
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methylpyridine
Hazard Category H227-H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H301-H311-H227-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only