missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylphenyl isothiocyanate, 98%

Catalog No. AAB2529614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2529614 25 g
AAB2529606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2529614 Supplier Thermo Scientific Chemicals Supplier No. B2529614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19241-38-4 | C8H6BrNS | 228.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19241-38-4
Molecular Formula C8H6BrNS
Molecular Weight (g/mol) 228.107
MDL Number MFCD00041087
InChI Key YASXCQRGYJGIKD-UHFFFAOYSA-N
Synonym 4-bromo-2-methylphenyl isothiocyanate, 4-bromo-1-isothiocyanato-o-toluene, 4-bromo-2-methylphenylisothiocyanate, 4-bromo-2-methyl-phenyl isothiocyanate, benzene, 4-bromo-1-isothiocyanato-2-methyl, acmc-1bu0e, #, benzene,4-bromo-1-isothiocyanato-2-methyl
PubChem CID 87977
IUPAC Name 4-bromo-1-isothiocyanato-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)N=C=S

Specifications

Melting Point 44°C to 46°C
Boiling Point 129°C to 130°C (3 mmHg)
Quantity 25 g
UN Number UN2923
Beilstein 2717248
Sensitivity Moisture sensitive
Formula Weight 228.12
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-methylphenyl isothiocyanate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 242-907-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only