missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylphenol, 97%

Catalog No. p-7031287
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3230706 5 g
AAH3230714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3230706 Supplier Thermo Scientific Chemicals Supplier No. H3230706

CAS: 2362-12-1 | C7H7BrO | 187.04 g/mol

4-Bromo-2-methylphenol, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-methylphenol, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 2362-12-1
MDL Number MFCD00055435

Specifications

Melting Point 62°C to 66°C
Odor Phenol-like
Molecular Formula C7H7BrO
Quantity 5 g
Synonym 4-bromo-o-cresol, 2-methyl-4-bromophenol, 4-bromo-2-methyl-phenol, o-cresol, 4-bromo, phenol, 4-bromo-2-methyl, 2-hydroxy-5-bromotoluene, p-bromocresol, 4-bromo-cresol, pubchem7106, acmc-1chy8
Solubility Information Slightly soluble in water.
InChI Key IWJGMJHAIUBWKT-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)Br)O
IUPAC Name 4-bromo-2-methylphenol
Molecular Weight (g/mol) 187.04
PubChem CID 16896
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methylphenol
Show More Show Less
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber GO6868000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only