missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylbiphenyl, 98%

Catalog No. p-7047179
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1787503 1 g
AAL1787506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1787503 Supplier Thermo Scientific Chemicals Supplier No. L1787503
Only null left

CAS: 5002-26-6 | C13H11Br | 247.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5002-26-6
Molecular Formula C13H11Br
Molecular Weight (g/mol) 247.135
MDL Number MFCD03093069
InChI Key ZBNARPVMXYNXQQ-UHFFFAOYSA-N
Synonym 4-bromo-2-methylbiphenyl, 4-bromo-2-methyl-1,1'-biphenyl, acmc-1asrj, 2-methyl-4-bromobiphenyl, 4-bromo-2-methyl-biphenyl, 3-methyl-4-phenylbromobenzene, 4-bromanyl-2-methyl-1-phenyl-benzene
PubChem CID 9964845
IUPAC Name 4-bromo-2-methyl-1-phenylbenzene
SMILES CC1=C(C=CC(=C1)Br)C2=CC=CC=C2

Specifications

Boiling Point 113°C to 114°C (0.7 mmHg)
Refractive Index 1.621
Quantity 1 g
Beilstein 1908937
Formula Weight 247.14
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-methylbiphenyl
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only