missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylbenzyl Alcohol 98.0+%, TCI America™

Catalog No. B52311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52311G 1 g
B52315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52311G Supplier TCI America Supplier No. B52311G

Chemical Identifiers

CAS 17100-58-2
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD11847398
InChI Key IFKWLKCPUIQXPU-UHFFFAOYSA-N
PubChem CID 22280088
IUPAC Name (4-bromo-2-methylphenyl)methanol
SMILES CC1=C(CO)C=CC(Br)=C1

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 1 g
Formula Weight 201.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-methylbenzyl Alcohol