missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylbenzenesulfonyl chloride, 97%

Catalog No. p-7030936
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3183509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3183509 Supplier Thermo Scientific Chemicals Supplier No. H3183509
Only null left

CAS: 139937-37-4 | C7H6BrClO2S | 269.537 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139937-37-4
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.537
MDL Number MFCD01365820
InChI Key KNBSFUSBZNMAKU-UHFFFAOYSA-N
Synonym 4-bromo-2-methylbenzene-1-sulfonyl chloride, 4-bromo-2-methylbenzenesulphonyl chloride, 4-bromo-2-methyl-benzenesulfonyl chloride, 2-methyl-4-bromobenzenesulfonylchloride, 5-bromo-2-chlorosulphonyl toluene, 2-methyl-4-bromobenzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-methyl, pubchem5066, buttpark 9857-56, intermediates-zcf02072
PubChem CID 2781338
IUPAC Name 4-bromo-2-methylbenzenesulfonyl chloride
SMILES CC1=C(C=CC(=C1)Br)S(=O)(=O)Cl

Specifications

Melting Point 63°C to 65°C
Boiling Point 164°C (15 mmHg)
Quantity 10 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 269.55
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methylbenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only