accessibility menu, dialog, popup

UserName

CH3BrSOOCl

CAS RN 139937-37-4

IUPAC Name: 4-bromo-2-methylbenzene-1-sulfonyl chloride
Synonyms: 4-bromo-2-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 269.54
Molecular Formula: C7H6BrClO2S
InChi Key: KNBSFUSBZNMAKU-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(Br)=C1)S(Cl)(=O)=O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)