missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 139937-37-4 |
|---|---|
| Molecular Formula | C7H6BrClO2S |
| Molecular Weight (g/mol) | 269.55 |
| InChI Key | KNBSFUSBZNMAKU-UHFFFAOYSA-N |
| Synonym | 4-bromo-2-methylbenzene-1-sulfonyl chloride, 4-bromo-2-methylbenzenesulphonyl chloride, 4-bromo-2-methyl-benzenesulfonyl chloride, 2-methyl-4-bromobenzenesulfonylchloride, 5-bromo-2-chlorosulphonyl toluene, 2-methyl-4-bromobenzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-methyl, pubchem5066, buttpark 9857-56, intermediates-zcf02072 |
| PubChem CID | 2781338 |
| IUPAC Name | 4-bromo-2-methylbenzenesulfonyl chloride |
| SMILES | CC1=C(C=CC(=C1)Br)S(=O)(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC451780100
|
N/A | 10g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 139937-37-4 | |
| 269.55 | |
| 4-bromo-2-methylbenzene-1-sulfonyl chloride, 4-bromo-2-methylbenzenesulphonyl chloride, 4-bromo-2-methyl-benzenesulfonyl chloride, 2-methyl-4-bromobenzenesulfonylchloride, 5-bromo-2-chlorosulphonyl toluene, 2-methyl-4-bromobenzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-methyl, pubchem5066, buttpark 9857-56, intermediates-zcf02072 | |
| 4-bromo-2-methylbenzenesulfonyl chloride |
| C7H6BrClO2S | |
| KNBSFUSBZNMAKU-UHFFFAOYSA-N | |
| 2781338 | |
| CC1=C(C=CC(=C1)Br)S(=O)(=O)Cl |
Specifications
| 139937-37-4 | |
| 96% min. (GC) | |
| Br(CH3)C6H3SO2Cl | |
| KNBSFUSBZNMAKU-UHFFFAOYSA-N | |
| 4-bromo-2-methylbenzenesulfonyl chloride | |
| 2781338 | |
| 97% |
| 164.0°C (15.0 mmHg) | |
| C7H6BrClO2S | |
| 4-bromo-2-methylbenzene-1-sulfonyl chloride, 4-bromo-2-methylbenzenesulphonyl chloride, 4-bromo-2-methyl-benzenesulfonyl chloride, 2-methyl-4-bromobenzenesulfonylchloride, 5-bromo-2-chlorosulphonyl toluene, 2-methyl-4-bromobenzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-methyl, pubchem5066, buttpark 9857-56, intermediates-zcf02072 | |
| CC1=C(C=CC(=C1)Br)S(=O)(=O)Cl | |
| 269.55 | |
| 269.55 | |
| 4-Bromo-2-methylbenzene-1-sulfonyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
RUO – Research Use Only