missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylbenzaldehyde, 95%

Catalog No. p-7053701
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61314MD 250 mg
AAH6131403 1 g
AAH6131406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH61314MD Supplier Thermo Scientific Chemicals Supplier No. H61314MD

CAS: 24078-12-4 | C8H7BrO | 199.047 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24078-12-4
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.047
MDL Number MFCD07787171
InChI Key RCBPVESMGNZMSG-UHFFFAOYSA-N
Synonym 4-bromo-2-methyl-benzaldehyde, 2-methyl-4-bromobenzaldehyde, 5-bromo-2-formyltoluene, benzaldehyde, 4-bromo-2-methyl, acmc-1chij, 4-brom-2-methylbenzaldehyd, ksc494k3j, 4-bromo-2-methyl benzaldehyde, benzaldehyde,4-bromo-2-methyl
PubChem CID 13555150
IUPAC Name 4-bromo-2-methylbenzaldehyde
SMILES CC1=C(C=CC(=C1)Br)C=O

Specifications

Quantity 250 mg
Formula Weight 199.04
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Bromo-2-methylbenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only