missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylanisole, 98+%

Catalog No. AAB2162109
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2162109 10 g
AAB2162118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2162109 Supplier Thermo Scientific Chemicals Supplier No. B2162109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14804-31-0 | C8H9BrO | 201.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14804-31-0
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD01321139
InChI Key UDLRGQOHGYWLCS-UHFFFAOYSA-N
Synonym 4-bromo-2-methylanisole, 5-bromo-2-methoxytoluene, benzene, 4-bromo-1-methoxy-2-methyl, 4-bromo-1-methoxy-2-methyl-benzene, 2-methyl-4-bromoanisole, 4-bromo2-methylanisole, acmc-1cfej, 4-bromo2-methyl-anisole, 4-bromo-2-methyl anisole, 4-bromo-2-methyl-anisole
PubChem CID 608315
IUPAC Name 4-bromo-1-methoxy-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)OC

Specifications

Melting Point 66°C to 69°C
Boiling Point 124°C to 125°C (25 mmHg)
Quantity 10 g
Beilstein 208224
Formula Weight 201.06
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-2-methylanisole
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only