missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylaniline, 98%

Catalog No. p-7022662
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1554314 25 g
AAA1554322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1554314 Supplier Thermo Scientific Chemicals Supplier No. A1554314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 583-75-5 | C7H8BrN | 186.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 583-75-5
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00007825
InChI Key PCHYYOCUCGCSBU-UHFFFAOYSA-N
Synonym 2-amino-5-bromotoluene, 4-bromo-o-toluidine, 2-methyl-4-bromoaniline, benzenamine, 4-bromo-2-methyl, o-toluidine, 4-bromo, 4-bromo-2-methyl-phenylamine, 4-bromo-2-methylbenzenamine, 5-bromo-2-aminotoluene, 1-amino-4-bromo-2-methylbenzene, 4-bromo-2-methylbenenamine
PubChem CID 11423
IUPAC Name 4-bromo-2-methylaniline
SMILES CC1=CC(Br)=CC=C1N

Specifications

Melting Point 56°C to 60°C
Boiling Point 240°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 25 g
UN Number UN2811
Beilstein 636521
Formula Weight 186.06
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-methylaniline
Hazard Category H315-H319-H335-H373
Hazard Statement GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 209-519-9
RTECSNumber XU4250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only