missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-methyl-1-indanone 98.0+%, TCI America™
$96.94 - $292.65
Chemical Identifiers
| CAS | 174702-59-1 |
|---|---|
| Molecular Formula | C10H9BrO |
| Molecular Weight (g/mol) | 225.085 |
| MDL Number | MFCD09881709 |
| InChI Key | AHMJTIYXKJRVFF-UHFFFAOYSA-N |
| PubChem CID | 10537152 |
| IUPAC Name | 4-bromo-2-methyl-2,3-dihydroinden-1-one |
| SMILES | CC1CC2=C(C1=O)C=CC=C2Br |
Chemical Identifiers
| 174702-59-1 | |
| 225.085 | |
| AHMJTIYXKJRVFF-UHFFFAOYSA-N | |
| 4-bromo-2-methyl-2,3-dihydroinden-1-one |
| C10H9BrO | |
| MFCD09881709 | |
| 10537152 | |
| CC1CC2=C(C1=O)C=CC=C2Br |
Specifications
| 174702-59-1 | |
| 134°C | |
| MFCD09881709 | |
| AHMJTIYXKJRVFF-UHFFFAOYSA-N | |
| 4-bromo-2-methyl-2,3-dihydroinden-1-one | |
| 10537152 | |
| ≥98.0% (GC) | |
| 4-Bromo-2-methyl-1-indanone |
| White-Yellow | |
| C10H9BrO | |
| 1 g | |
| CC1CC2=C(C1=O)C=CC=C2Br | |
| 225.085 | |
| 225.09 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No